CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.175" Diameter
Dodavatel: Avantor Fluid Handling
Mix, react, and/or separate liquid-liquid mixtures in a single device.
- Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
- Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.