Tisk…
CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.100" Diameter
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX17315-21
Your Organization's Price:
Mix, react, and/or separate liquid-liquid mixtures in a single device.
  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.