Tisk…
Tygoprene Transfer Tubing, Tygoprene® XL-60, 1/8" ID x 1/4" OD; 50 Ft
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX95870-02
Your Organization's Price:
Use in varying temperatures.
  • FDA compliant for food contact
<B>Formulation:</B> Tygoprene XL-60<br /><B>Applications:</B> Ink transfer, soap and detergent delivery, cold or hot beverage transfer and dispenseing, food processing, general chemical transfer and processing.<br /><B>Characteristics:</B> Low extractables, excellent resistance to ozone, oxygen, and sunlight. Resists a wide range of fluids, including acids and bases. Translucent.<br /><B>Certification:</B> FDA compliant. Tubing is NSF listed under standard 51 and is UL 94-HB flammability resistant.<br /><B>Temperature range:</B> — 87 to 250 °F ( — 66 to 121 °C)<br /><B>Sterilize:</B> Autoclave, ethylene oxide, or gamma irradiation.