Tisk…
Masterflex® L/S® Easy-Load® II Pump Head Replacement Rotor Assembly for High-Performance Precision Tubing, SS
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX77200-69
Your Organization's Price:
Get the most our of your Masterflex® L/S® Easy-Load® II Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® pump head
  • Suitable for use with Easy-Load® II pump head 77200-62
Replace your pump head rotor assembly to extend the life of your pump head.