Masterflex® L/S® Easy-Load® II Pump Head Replacement Rotor Assembly for High-Performance Precision Tubing, CRS
Dodavatel: Avantor Fluid Handling
Get the most our of your Masterflex® L/S® Easy-Load® II Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® pump head
- Suitable for use with Easy-Load® II pump head 77200-52
Replace your pump head rotor assembly to extend the life of your pump head.