Tisk…
Masterflex® Cable Assembly for Remote Control; DB25(M) Connector; 25 Ft
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX07523-95
Your Organization's Price:
Easily connect your Masterflex® drive to a PLC or other control device.
  • Constructed of durable PVC
  • Long 25 ft (7.9 m) cable length makes connection simple
  • Cable features 26 AWG wire gauge
  • Connection Type: DB25(M)
Cable assemblies include terminal connector and 25 ft (7.9 m) cable with stripped wire ends. Verify the connector type on your drive before ordering