Tisk…
Catalyst Stopped Tubing, Platinum-Cured Silicone, 2.79 mm ID; 6/Pk
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX97200-08
Your Organization's Price:
Start pumping with little effort - preconfigured Links for quick and easy setup.
  • Four formulations offer wide range of chemical compatibility
  • Save time with already assembled links
  • Use with the Catalyst extenstion coils / tubing to complete your system
  • FH15 and FH30 utilize the stopped assemblies that have an ID of: 0.8, 1.6, 3.2, and 4.8 mm
  • FH100M (with the large cartridges) can use assemblies with an ID of: 0.8, 1.6, 3.2, 4.8, and 6.4 mm
  • FH100M (with the small cartridges) can use assemblies with an ID of: 0.16, 0.25, 0.89, 1.42, 2.06, and 2.79 mm
To complete your Catalyst® pumping system, order two-stop color-coded and extension tubing. Tubing comes in various formulations for almost any pumping application.<br /><br />The stops on the two-stop tubing are color-coded according to tubing ID. <br /><br />Order extension tubing separately. For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.