Tisk…
Masterflex® Transfer Tubing, C-Flex®, Clear, 1/4" ID x 3/8" OD; 25 Ft
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX06422-10
Your Organization's Price:
Meets the demands of various industries.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Tubing size: 1/4" ID x 3/8" OD
Applications: Pharmaceutical, biotech, and general-purpose lab uses requiring high performance. Economical alternative to platinum silicone.<br />Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Transparent.<br />Certification: Product is manufactured in accordance with Part 820 Quality System Requirements (GMP); and meets USP Class VI.<br />Temperature range: -40 to 275 °F (-40 to 135 °C)<br />Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
See 96124-16 for the irradiated version of this tubing.