BarbLock Assembly Hand Tool for 1/2" and I/P 73 Tubing
Dodavatel: Avantor Fluid Handling
Easily install your Barblock Fittings.
- Optimize performance of your Barblock fittings
Select a hand tool to assemble your BarbLock Retainers based on the tubing size. Universal removal tool is also available.
<b>Informace o dodávce: </b>10” plier for the BarbLock hand tools.