Tisk…
Masterflex® B/T® PerfectPosition® Pump Tubing, PharMed® BPT, B/T 87; 10 ft
Dodavatel: Avantor Fluid Handling
VWR Item#MFLX06507-87
Your Organization's Price:
Ensure optimum performance from your Masterflex® pump.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Up to 10,000 hours of tubing life
  • Resists ozone and UV radiation
  • Noncytotoxic and nonhemolytic
  • Ideal for tissue and cell culture work
  • Heat sealable and bondable
  • Opaque to UV and visible light to protect light-sensitive fluids
  • Low gas permeability
  • High-pressure 103 bar (150 psi) version available (L/S High Pressure family)
  • PerfectPosition™ tubing retention marks indicate the exact placement of tubing in the pump head to provide the best performance and life of the tubing
  • Long service life minimises risk of fluid exposure; reduces tubing costs and pump downtime
Application suitability: Suitable for use with Acids, Alkalis, Pressure, Vacuum, Viscous fluid, and Sterile Fluids. Not Recommended for use with organic solvents.<br /><br />Physical characteristics : Thermoplastic elastomer, polypropylene-based material with USP mineral oil. Excellent tensile strength, firm (stiff) material. Opaque, beige.<br /><br />Temperature range: <br />(static): -51 to 132C (-60 to 270F)<br />(dynamic): -20 to 100C (-4 to 212F)<br /><br />Meets classifications: <br />USP Class VI<br />European Pharmacopoeia (EP 3.2.9)<br /><br />Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples.<br /><br />Sterilize: EtO, Autoclave, Gamma Radiation