Tisk…
Disposable particle filtering respirators, FFP2/FFP3, Standard Plus-programm, zetMask®
Dodavatel: Zellstoff-Vertriebs-GmbH & Co. KG
30931-94
ZVGA30931-94EA 1590 CZK
ZVGA30931-94
Disposable particle filtering respirators, FFP2/FFP3, Standard Plus-programm, zetMask®
Polomasky a filtrační respirátory Polomasky filtrační, jednorázové Polomasky k filtraci částic
Ultralight, pre-shaped fine-dust filter masks with exhalation valve for protection against coarse, hazardous particle.
  • High degree of efficiency with minimum space requirements
  • Foam-padded nose area for pressure-free sealed fit
  • Elastic bands
  • Hygienically individually packed; in dispenser box
EN 149:2001
Certifikace: EN 149:2001