Tisk…
Ion selective electrode system, VARiON® Plus 700 IQ
Dodavatel: WTW ONLINE
107040
WTWO107040EA 168200 CZK
WTWO107040
Ion selective electrode system, VARiON® Plus 700 IQ
Elektrody Elektrody ISE
In-situ combination sensor for direct measurements of ammonium and nitrate using only one sensor. Designed for the installation of a measuring electrode, a reference electrode and a compensation electrode.
  • Automatic compensation of interferences
  • Integrated NTC thermistor
  • Calibration-free, long time stable
  • Display shows the already compensated values, the potassium measuring value can be displayed optionally