Tisk…
Hybridisation bottles
Dodavatel: witeg Labortechnik
2.446.115
WITG2.446.115EA 1370 CZK
WITG2.446.115
Hybridisation bottles
Láhve Láhve hybridizační
Borosilicate glass 3.3, with GL 45 screw cap and silicone sealing.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes