Tisk…
Dropping funnels, cylindrical, with pressure balance
Dodavatel: witeg Labortechnik
1.866.002
WITG1.866.002EA 5210 CZK
WITG1.866.002
Dropping funnels, cylindrical, with pressure balance
Nálevky Nálevky přikapávací
Dropping funnels, cylindrical, stopcock with PTFE key and screw-thread retaling nut.
  • Graduated
  • Excellent chemical resistance
  • Excellent temperature resistance