Tisk…
Distilling bridges
Dodavatel: witeg Labortechnik
1.205.150 1.205.260 1.205.300 1.205.301
WITG1.205.150EA 3250 CZK
WITG1.205.150 WITG1.205.260 WITG1.205.300 WITG1.205.301
Distilling bridges
Komponenty pro destilaci Nástavce
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.