Tisk…
Flasks, Erlenmeyer, with standard ground joints
Dodavatel: witeg Labortechnik
0.620.250 0.624.200 0.624.002 0.624.300 0.624.003 0.624.005 0.619.010.BR 0.619.025.BR 0.619.005.BR 0.620.025 0.620.050 0.622.250.BR 0.622.500.BR 0.622.100.BR 0.620.100 0.620.005 0.622.001.BR 0.622.002.BR 0.624.001.BR 0.620.025.BR 0.620.250.BR 0.620.100.BR 0.620.050.BR 0.621.050.E 0.621.100.E 0.621.200.E 0.621.250.E 0.621.300.E 0.621.500.E 0.621.001.E 0.621.002.E 0.622.001.E 0.622.002.E 0.624.250.E 0.624.300.E 0.624.500.E 0.624.001.E 0.624.002.E 0.620.100.E 0.620.250.E 0.619.005.E 0.619.010.E 0.619.025.E 0.619.050.E 0.619.100.E 0.619.250.E 0.620.005.E 0.620.010.E 0.620.025.E 0.620.050.E 0.622.050.E 0.622.100.E 0.622.200.E 0.622.250.E 0.622.300.E 0.622.500.E
201-0184EA 393 CZK
201-0184 201-0188 WITG0.624.002 201-0189 201-0190 201-0196 WITG0.619.010.BR WITG0.619.025.BR WITG0.619.005.BR WITG0.620.025 WITG0.620.050 WITG0.622.250.BR WITG0.622.500.BR WITG0.622.100.BR WITG0.620.100 201-0181 WITG0.622.001.BR WITG0.622.002.BR WITG0.624.001.BR WITG0.620.025.BR WITG0.620.250.BR WITG0.620.100.BR WITG0.620.050.BR WITG0.621.050.E WITG0.621.100.E WITG0.621.200.E WITG0.621.250.E WITG0.621.300.E WITG0.621.500.E WITG0.621.001.E WITG0.621.002.E WITG0.622.001.E WITG0.622.002.E WITG0.624.250.E WITG0.624.300.E WITG0.624.500.E WITG0.624.001.E WITG0.624.002.E WITG0.620.100.E WITG0.620.250.E WITG0.619.005.E WITG0.619.010.E WITG0.619.025.E WITG0.619.050.E WITG0.619.100.E WITG0.619.250.E WITG0.620.005.E WITG0.620.010.E WITG0.620.025.E WITG0.620.050.E WITG0.622.050.E WITG0.622.100.E WITG0.622.200.E WITG0.622.250.E WITG0.622.300.E WITG0.622.500.E
Flasks, Erlenmeyer, with standard ground joints
Baňky Baňky dle Erlenmeyera
DURAN®, borosilicate glass 3.3 or borosilicate glass 3.3 (Economy type), clear or amber.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Volume imprint.