Tisk…
Screw caps for thread connection
Dodavatel: witeg Labortechnik
0.176.032
WITG0.176.032EA 95 CZK
WITG0.176.032
Screw caps for thread connection
Uzávěry Uzávěry k láhvím
PBT, red without seals.
  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable, sterilisable with hot air
With central hole.