Tisk…
Burette, straight, class A, PTFE stopcock, SP Wilmad-LabGlass
Dodavatel: SP Industries
LG-4011T-102
WILMLG-4011T-102EA 22190 CZK
WILMLG-4011T-102
Burette, straight, class A, PTFE stopcock, SP Wilmad-LabGlass
Byrety
Burette, made of borosilicate glass, with funnel shaped top to accommodate a rubber stopper and glass tubing to fill from a delivery tip via suction.
  • Precision type burette
  • With durable coloured scale
  • Stopcock with 2 mm precision bare PTFE plug