Tisk…
Oil sample bottle, with cork, WHEATON®
Dodavatel: DWK Life Sciences
W216994
WHEAW216994EA 9640 CZK
WHEAW216994
Oil sample bottle, with cork, WHEATON®
Láhve Láhve skleněné
Clear, Type III soda-lime glass.
Bottles come with cork inserted.