Tisk…
Culture shake flasks, WHEATON®
Dodavatel: DWK Life Sciences
354235 354237 354239 354242 354244 353255 353257 353259 353262 353264 353266
WHEA354235EA 1900 CZK
WHEA354235 734-1399 734-1400 WHEA354242 WHEA354244 WHEA353255 WHEA353257 WHEA353259 WHEA353262 WHEA353264 WHEA353266
Culture shake flasks, WHEATON®
Baňky Baňky kultivační
These culture shake flasks are made of 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
  • Baffles improve aeration and mixing for microbial and bacterial cultures
  • Break resistant lip for use with stopper, cotton plug or film cover
  • Pinched neck design to reduce splashing on flasks with four baffles
Designed for use with orbital or reciprocating shakers.