Tisk…
pH Indicator papers, in Books, Whatman™
Dodavatel: Whatman products (Cytiva)
2600-500
WHAT2600-500EA 3810 CZK
WHAT2600-500
pH Indicator papers, in Books, Whatman™
Papírky testovací Stripy pH indikační
These pH indicators have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparison table supplied allows the pH to be determined.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
The book format is particularly suitable for educational and industrial use. In schools this format is economical because the amount of paper per student can be carefully controlled.