Tisk…

Benzoan sodný 99.0-100.5% Ph. Eur.

Dodavatel: VWR Chemicals
Varování

Synonyms: Benzoic acid sodium salt

83551.290EA 2250 CZK
83551.290 83551.360 VWRC83551.461
Benzoan sodný 99.0-100.5% Ph. Eur.
Benzoan sodný
Vzorec: C₆H₅COONa
MW: 144.11
Bod tání: 410…430 °C
Hustota: 1.44
Bod vzplanutí: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS číslo: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Objednat

Výsledky specifikačního testu

Assay (calculated on dried substance) 99.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Appearance of solution Passes test
Acidity or alkalinity Passes test
Halogenated compounds Passes test
Heavy metals (as Pb) Max. 10 ppm
Loss on drying (105°C) Max. 2 %
Statement of original manufacturer (*)
Benzene (*) Max. 2 ppm
Toluene (*) Max. 890 ppm
Other residual solvents (*) Unlikely by manuf.process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR