Tisk…

n-Heptan 99,0, AnalaR NORMAPUR® zA

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: high purity solvents

24551.290EA 2560 CZK
24551.290 24551.324 24551.368 24551.461 VWRC24551.550
n-Heptan 99,0, AnalaR NORMAPUR® zA
n-Heptane
Vzorec: H₃C(CH₂)₅CH₃
MW: 100.2
Bod varu: 98.4 °C (1013 hPa)
Bod tání: –90,6 °C
Hustota: 0.6838 g/cm³ (20 °C)
Bod vzplanutí: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS číslo: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
REACH: 01-2119457603-38
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay Min. 99.0 %
IR Spectrum Passes test
Acidity Max. 0.0001 meq/g
Alkalinity Max. 0.0001 meq/g
Boiling point 98 - 99 °C
Bromine value Max. 0.5
Colouration Max. 10 APHA
Density (20/4) 0.681 - 0.685
Substances coloured by H2SO4 Max. 90 APHA
Aromatics (V/V) Max. 0.1 %
Evaporation residue Max. 10 ppm
Total S (as SO4) Max. 10 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.1 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.02 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR