Tisk…
Arium® Mini Xtend Ultrapure Water Systems
Dodavatel: Sartorius
H2O-MAR-T H2O-MAR-UV-T H2O-MUR-T H2O-MUR-UV-T
VIVAH2O-MAR-TEA 175850 CZK
VIVAH2O-MAR-T VIVAH2O-MAR-UV-T VIVAH2O-MUR-T VIVAH2O-MUR-UV-T
Arium® Mini Xtend Ultrapure Water Systems
Systémy pro výrobu čisté vody Systémy pro výrobu čisté a ultračisté vody
Compact Arium® Mini laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 L per day and are ideal for use in preparation of buffers, media and samples, both in life sciences applications and in analytical laboratory procedures.
  • Flexible: Different versions for different inlet water and connection options, as well as the desired water withdrawal
  • Compact: Space-saving with a width of only 28 cm (+9 cm for versions with attached dispensing unit)
  • Intuitive: Color touch display, with direct access to all important
More Product Information