Tisk…
SCHOTT Tubular Glass Injection Vials
Dodavatel: Lokální dodavatel
VCDIN20R
VITUVCDIN20REA 46360 CZK
VITUVCDIN20R
SCHOTT Tubular Glass Injection Vials
Vialky
Clear and amber neutral Type I/Type 1 tubular glass vials and lyophilisation (Lyo) vials.
  • Glass vials produced by SCHOTT using SCHOTT Fiolax® neutral Type I glass tubing
  • Standard 13 mm and 20 mm crimp neck finishes
  • 100% camera inspection of dimensional parameters
  • Vials compliant with EP, JP and USP
Certifikace: Manufactured and packed according to ISO 9001 and ISO 15378.