Tisk…
Absorbent mats, SpillEx
Dodavatel: FHP VILEDA SCS
146760
VILE146760EA 2010 CZK
VILE146760
Absorbent mats, SpillEx
Ochrana povrchu Papíry ochranné na pracovní desky
Disposable absorption mats are designed to absorb water-based liquids very quickly.
  • Single use, absorbs up to 1200 ml of water-based liquid
  • Absorbent polymers turn into a gel in contact with liquids