Tisk…
Foam squeegee
Dodavatel: FHP VILEDA SCS
141972 141973
VILE141972EA 819 CZK
VILE141972 VILE141973
Foam squeegee
Mopy
Professional foam blade squeegees for superior and ergonomic dragging results. Ideal for places where large amounts of water must be dispersed. Hygiene foam squeegee is recommended, when hygiene is extremely important. For wet push and pull applications.
  • Hygienic foam with closed cells (high density)
  • Color coded caps (changeable for each work zone, minimising the risk of cross-contamination)
  • Splash guard, at both sides of the frame, to prevent the liquid from spilling over the squeegee
Note: The handles that fix the squeegee VILE141973 (60 cm) are 129-More Product Information