Tisk…
Cleaning trolley system, Origo
Dodavatel: FHP VILEDA SCS
121560 121594
129-0850EA 20040 CZK
129-0850 VILE121594
Cleaning trolley system, Origo
Vozíky
The Origo trolley system has been designed for maximum flexibility and changeability. The user can either choose one of the standard configurations or have the trolley customised exactly to his requirements, and when requirements change so the system can be reconfigured. The 300 series offer the abitility to retro-fit and re-customise the trolley over and over, offering almost limitless possibilities.
  • Adjustable handlebars
  • Height adjustable trays and drawers
  • Wide choice of castor size great manoeuvrability and thought-through design result in a state-of-
More Product Information