Tisk…
Tubing, PTFE, Chemfluor®
Dodavatel: Saint Gobain Life Sciences
TFE0213050 TFE0423050 TFEBZC4025 TFEA4C4100 TFE0413050 TFE0519050 TFE1013050 TFEAUBP100 TFEAUC4025 TFEBEC4025 TFEBEC4050 TFEBPBP050 TFEC4C4025 TFECJC4025 TFECJC4100 TFEDPC4100
228-1054EA 0 CZK
228-1054 228-1709 VERNTFEBZC4025 VERNTFEA4C4100 VERNTFE0413050 VERNTFE0519050 VERNTFE1013050 VERNTFEAUBP100 VERNTFEAUC4025 VERNTFEBEC4025 VERNTFEBEC4050 VERNTFEBPBP050 VERNTFEC4C4025 VERNTFECJC4025 VERNTFECJC4100 VERNTFEDPC4100
Tubing, PTFE, Chemfluor®
Hadičky Přenosová hadička
PTFE, opaque white to translucent in appearance.
  • Fully fluorinated fluoropolymer tubing
  • High degree of rigidity
  • Very good chemical resistance
Produces smooth, non-porous, dimensionally stable tubing made out of polytetrafluoroethylene resin, PTFE tubing has the highest working temperature. It is widely used as a pressure tubing in general chemical applications such as delivery of natural gas and mineral oils, toxic gas monitoring and paint, varnish or adhesive delivery lines. PTFE tubing also services numerous applications in the automotive,More Product Information