Tisk…
Tygon® A-60-C Tubings
Dodavatel: Saint Gobain Life Sciences
AZT00003-S AZT00004-S AZT00007-S AZT00008-S AZT00009-S AZT00010-S AZT00012-S AZT00017-S AZT00018-S AZT00019-S AZT00022-S AZT00023-S AZT00027-S AZT00028-S AZT00029-S AZT00038-S AZT00046-S AZT00053-S
VERNAZT00003-SEA 5520 CZK
VERNAZT00003-S VERNAZT00004-S VERNAZT00007-S VERNAZT00008-S VERNAZT00009-S VERNAZT00010-S VERNAZT00012-S VERNAZT00017-S VERNAZT00018-S VERNAZT00019-S VERNAZT00022-S VERNAZT00023-S VERNAZT00027-S VERNAZT00028-S VERNAZT00029-S VERNAZT00038-S VERNAZT00046-S VERNAZT00053-S
Tygon® A-60-C Tubings
Hadičky Přenosová hadička
Tygon® A-60-C tubing from Saint-Gobain is formulated to withstand extreme temperatures from −59 to 135 °C (−75 to 275 °F) and will not crack or deteriorate when used in demanding cleaning chemical dispensing applications. Extremely flexible, it resists kinks and retains its shape while installing quickly and easily.
  • Long flex life in peristaltic pumps
  • Compatible with virtually all common sanitisers and cleaners
  • Ozone and UV light resistant
  • Repeatedly autoclavable
  • Chemically compatible with a wide range of food types and cleaners
Tygon® A-60-C tubingMore Product Information
New Product
Objednat
Produktová specifikace
Environmentally Preferable
Maximální teplota 135 °C
Minimální teplota −60 °C