Tisk…
Tubing, Tygon® A-60-G
Dodavatel: Saint Gobain Life Sciences
AFL00007-S AFL00027 AFL00012-S AFL00018-S AFL00029-S AFL00019-S AFL00003-S AFL00023-S AFL00008-S AFL00010-S AFL00013-S AFL00015-S AFL00017-S AFL00020-S AFL00026-S AFL00027-S AFL00028-S AFL00032-S AFL00036-S AFL00037-S AFL00038-S AFL00045-S AFL00046-S AFL00053-S
VERNAFL00007-SEA 3020 CZK
VERNAFL00007-S VERNAFL00027 VERNAFL00012-S VERNAFL00018-S VERNAFL00029-S VERNAFL00019-S VERNAFL00003-S VERNAFL00023-S VERNAFL00008-S VERNAFL00010-S VERNAFL00013-S VERNAFL00015-S VERNAFL00017-S VERNAFL00020-S VERNAFL00026-S VERNAFL00027-S VERNAFL00028-S VERNAFL00032-S VERNAFL00036-S VERNAFL00037-S VERNAFL00038-S VERNAFL00045-S VERNAFL00046-S VERNAFL00053-S
Tubing, Tygon® A-60-G
Hadičky Přenosová hadička
Thermoplastic elastomer, black.
  • Long lasting, weatherproof, good abrasion resistance and exceptional resistance to fatigue caused by continual flexing
  • Excellent chemical resistance
  • Low gas permeability compared with rubber tubing
Versatile tubing for industrial and laboratory applications. Heat- and ozone-resistant. Excellent resistance to inorganic liquids (acids and bases). Ideal for use in peristaltic pumps.
Temperature resistance: −60 to +135 °C
Hardness (Shore durometer A): 61
Density: 0,98