Tisk…
Pump tubing, Tygon® E-1000
Dodavatel: Saint Gobain Life Sciences
ADK00003 ADK00007 ADK00012 ADK00017 ADK00019 ADK00022 ADK00027 ADK00029 ADK00036 ADK00038
VERNADK00003EA 5210 CZK
VERNADK00003 VERNADK00007 VERNADK00012 VERNADK00017 VERNADK00019 VERNADK00022 VERNADK00027 VERNADK00029 VERNADK00036 VERNADK00038
Pump tubing, Tygon® E-1000
Hadičky Přenosová hadička
Tygon®, translucent.
  • Soft and flexible
  • Low-temperature resistant for low level cryogenic applications
  • Stays flexible at temperatures as low as –55 °C
  • Resistant to corrosive chemicals for laboratory applications
  • Low durometer for use in low-torque pump applications
Tygon® E-1000 tubing is an excellent solution to applications requiring complicated laboratory set-ups with sharp radius curves and multiple directional changes. This tubing resists twisting and collapse, which are common problems when using other tubing products. This tubing deliversMore Product Information