Tisk…
Tubing, silicone, Versilon® SPX-50
Dodavatel: Saint Gobain Life Sciences
ABX00002 ABX00004 ABX00007 ABX00017 ABX00053 ABX00003
VERNABX00002EA 0 CZK
VERNABX00002 VERNABX00004 VERNABX00007 VERNABX00017 VERNABX00053 VERNABX00003
Tubing, silicone, Versilon® SPX-50
Hadičky Přenosová hadička
Silicone, peroxide-cured, translucent.
  • Provides resiliency, long life and durability
  • Ultra-smooth inner bore reduces risk of particulate entrapment
  • Taste and odour free
  • Non-toxic, non-hemolytic and non-pyrogenic
These high-strength silicone tubing produced from a proprietary combination of silicone elastomers, provides durability and versatility in a range of applications. It optimizes critical physical properties such as tensile strength, elongation and compression set, resulting in a more physically durable product. Ideal for veterinary, medical andMore Product Information