Tisk…
Tubing, silicone, Versitec®
Dodavatel: Saint Gobain Life Sciences
761839 761830 761857
VERN761839EA 3930 CZK
VERN761839 VERN761830 VERN761857
Tubing, silicone, Versitec®
Hadičky Přenosová hadička
Silicone, peroxide cured, translucent.
  • Water-repellent
  • Anti-adhesive
  • Exceptional resistance to UV rays, oxygen and ozone
  • Ppermeable.
Versitec tubing is a high quality industrial silicone used in food industry, household electrical appliances, electrical industry and in drink dispensers. The technical performance of the Versitec® tubing, together with its effectiveness make it the ideal means of transferring high purity or high-tech fluids.
Temperature resistance: -60 to +200 °C
Hardness (Shore durometer A): 40
Density: 1,15
Certifikace: More Product Information