Tisk…
Stoppers, rubber, conical, pre-drilled
Dodavatel: Saint Gobain Life Sciences
203241
VERN203241EA 4570 CZK
VERN203241
Stoppers, rubber, conical, pre-drilled
Zátky Zátky kaučukové (gumové)
Natural rubber, grey-blue.
  • Resistant to common laboratory chemicals
  • Provide a perfect seal and exceptional bonding
  • With pre-drilled holes
  • Resistant to temperatures from −5 to +70 °C (+120 °C in humid atmosphere)
Highly elastic, for perfect sealing and adhesion.
Resistant to temperatures from -5 to +70 °C (120 °C in moist heat)
DIN 12871
Certifikace: DIN 12871.