Tisk…
Solid phase microextraction fibers
Dodavatel: VARIAN
SU57343U 391896303 391896314 391896315 391896316 SU57320U SU57341U SU57321U SU57323U SU57324U SU57325U SU57327U SU57328U SU57329U SU57335U SU57344U SU57345U SU57346U SU57348U SU57356U SU57357U 391896307 391896308 391896309 391896313 392548402 SU57347U SU57550U SU57551U 392609902 SU57354U SU57355U
VARISU57343UEA 0 CZK
VARISU57343U VARI391896303 VARI391896314 VARI391896315 VARI391896316 VARISU57320U VARISU57341U VARISU57321U VARISU57323U VARISU57324U VARISU57325U VARISU57327U VARISU57328U VARISU57329U VARISU57335U VARISU57344U VARISU57345U VARISU57346U VARISU57348U VARISU57356U VARISU57357U VARI391896307 VARI391896308 VARI391896309 VARI391896313 VARI392548402 VARISU57347U VARISU57550U VARISU57551U VARI392609902 VARISU57354U VARISU57355U
Solid phase microextraction fibers
Chromatografické potřeby Chromatography Spare Parts
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.