Tisk…
Polystyrene standards, GPC/SEC columns calibration
Dodavatel: VARIAN
PL2010-0102 PL2010-0104 PL2010-0105 PL2012-0001 PL2012-0005 PL2012-1005 PL2012-1010 PL2012-3001 PL2012-4001 PL2012-4005 PL2012-4010 PL2012-5001 PL2012-5005 PL2012-5010 PL2012-6001 PL2012-6005 PL2012-6010 PL2012-7001 PL2012-7005 PL2012-7010 PL2012-8001 PL2012-8005 PL2012-8010 PL2012-9001 PL2012-9005 PL2012-9010 PL2013-0001 PL2013-1001 PL2013-1005 PL2013-1010 PL2013-2001 PL2013-2005 PL2013-2010 PL2013-3001 PL2013-3005 PL2013-3010 PL2013-4001 PL2013-4005 PL2013-4010 PL2013-5001 PL2013-5005 PL2013-5010 PL2013-6001 PL2013-7001 PL2013-7010 PL2013-8001 PL2013-8005 PL2013-9001 PL2013-9005 PL2013-9010 PL2014-0001 PL2014-0005 PL2014-0010 PL2014-1001 PL2014-1005 PL2014-2001 PL2014-3001 PL2014-3005 PL2014-4001 PL2014-5001 PL2014-6001 PL2014-6005 PL2014-7001 PL2014-7005 PL2014-8001 PL2014-8005 PL2014-9001
VARIPL2010-0102EA 30320 CZK
VARIPL2010-0102 VARIPL2010-0104 VARIPL2010-0105 VARIPL2012-0001 VARIPL2012-0005 VARIPL2012-1005 VARIPL2012-1010 VARIPL2012-3001 VARIPL2012-4001 VARIPL2012-4005 VARIPL2012-4010 VARIPL2012-5001 VARIPL2012-5005 VARIPL2012-5010 VARIPL2012-6001 VARIPL2012-6005 VARIPL2012-6010 VARIPL2012-7001 VARIPL2012-7005 VARIPL2012-7010 VARIPL2012-8001 VARIPL2012-8005 VARIPL2012-8010 VARIPL2012-9001 VARIPL2012-9005 VARIPL2012-9010 VARIPL2013-0001 VARIPL2013-1001 VARIPL2013-1005 VARIPL2013-1010 VARIPL2013-2001 VARIPL2013-2005 VARIPL2013-2010 VARIPL2013-3001 VARIPL2013-3005 VARIPL2013-3010 VARIPL2013-4001 VARIPL2013-4005 VARIPL2013-4010 VARIPL2013-5001 VARIPL2013-5005 VARIPL2013-5010 VARIPL2013-6001 VARIPL2013-7001 VARIPL2013-7010 VARIPL2013-8001 VARIPL2013-8005 VARIPL2013-9001 VARIPL2013-9005 VARIPL2013-9010 VARIPL2014-0001 VARIPL2014-0005 VARIPL2014-0010 VARIPL2014-1001 VARIPL2014-1005 VARIPL2014-2001 VARIPL2014-3001 VARIPL2014-3005 VARIPL2014-4001 VARIPL2014-5001 VARIPL2014-6001 VARIPL2014-6005 VARIPL2014-7001 VARIPL2014-7005 VARIPL2014-8001 VARIPL2014-8005 VARIPL2014-9001
Polystyrene standards, GPC/SEC columns calibration
Standardy Standardy pro chromatografii
Polystyrene standards for many organic solvents either for conventional GPC column calibration or for calibrating light scattering and viscosity detectors that provide equidistant calibration points for greater accuracy.
  • Compatible with most organic solvents
  • Certificate of analysis meets international protocols
  • Calibration capability for virtually all applications