Tisk…
Micro pipette tips, OMIX®
Dodavatel: VARIAN
A57003100K A5700310K A57009100K A57003MB A57003MBK A5700910 A57009100 A5700910K A5700410 A57004100 A5700310 A57003100
VARIA57003100KEA 65630 CZK
VARIA57003100K VARIA5700310K VARIA57009100K VARIA57003MB VARIA57003MBK VARIA5700910 VARIA57009100 VARIA5700910K VARIA5700410 VARIA57004100 VARIA5700310 VARIA57003100
Micro pipette tips, OMIX®
Špičky pipetovací
Bond Elut OMIX pipette tips reliably purify and enrich femtomole and picomole levels of peptides and proteins prior to MALDI-TOF or LC/MS/MS. The unique monolithic sorbent technology used in Bond Elut OMIX consistently outperforms other tips by delivering uniform flow and strong analyte-to-surface interactions. Bond Elut OMIX tips bind more peptides to deliver better sequence coverage.
  • Fast, uniform flow maximizes productivity and reproducibility
  • Minimal peptide losses lead to higher recoveries
  • Available in three phases and sizes to deliver better sequence coverage