Tisk…
Flame atomisation systems
Dodavatel: VARIAN
9910024700 9910043700 9910044000 9910053300 9910053900 110634490 110637000 1610117390 210164000 210164100 190034100
VARI9910024700EA 1870 CZK
VARI9910024700 VARI9910043700 VARI9910044000 VARI9910053300 VARI9910053900 VARI110634490 VARI110637000 VARI1610117390 VARI210164000 VARI210164100 VARI190034100
Flame atomisation systems
Spektrofotometry a fotometry Fotometry plamenové
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tuneMore Product Information