Tisk…

Tetrabutylammonium hydroxide 0.4 M in aqueous solution for HPLC, for ion pair chromatography, J.T.Baker®

Dodavatel: Avantor

V365-07EA 15340 CZK
V365-07
Tetrabutylammonium hydroxide 0.4 M in aqueous solution for HPLC, for ion pair chromatography, J.T.Baker®
Tetrabutylammonium hydroxide
Vzorec: C₁₆H₃₇NO
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS číslo: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Objednat

Výsledky specifikačního testu

For Laboratory,Research,or Manufacturing Use 0.37 - 0.43
Molarity 0.37 - 0.43
Appearance Passes Test
Absorbance at 420 nm (neat) ≤0.150
Bromide (Br) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR