Tisk…
UV Meters, analog, J-221 & J-225
Dodavatel: UVP ULTRA VIOLET PRODUCTS
97-0004-01 97-0003-01
UVPA97-0004-01EA 30120 CZK
UVPA97-0004-01 UVPA97-0003-01
UV Meters, analog, J-221 & J-225
Expozimetry
The J-Series UV meters are photovoltaic devices for taking repeatable readings of UV light sources in either the 365 nm or 254 nm UV range.
  • Assures accurate readings
  • Self-powered for maximum portability
  • 5X attenuation screen and for remote readings, a four foot (1.2 m) cord connects the sensor to the meter
Certifikace: J-221 Meter complies with MIL STD 45662-A and can be found in MIL-I-6868D (NSN 6635-488-5451).
Informace o dodávce: Each meter comes with a sensor, attenuation screen and filter.