Tisk…
High visibility bump caps, uvex u-cap hi-viz
Dodavatel: UVEX
9794.800
UVEX9794.800EA 975 CZK
UVEX9794.800
High visibility bump caps, uvex u-cap hi-viz
Přilby Čepice s kšiltem
These premium caps in bright signal colours feature an innovative hard shell element for reliable impact protection.
  • Protects against abrasions and impact injuries
  • Integrated reflective piping on the brim and textile components
  • Flexible plastic shell adapts to different head shapes and sizes
  • Size adjustment can be done using adjustable band
  • With a long peak for increased sun protection
*One size fits all.
Certifikace: EN 471, EN 812