Tisk…
Safety helmets, uvex airwing B-S
Dodavatel: UVEX
9762.521 9762.421
UVEX9762.521EA 573 CZK
UVEX9762.521 UVEX9762.421
Safety helmets, uvex airwing B-S
Přilby Přilby bezpečnostní
Polyethylene helmet shell with three variable air vents and conventional suspension harness for a comfortable fit.
  • Shorter brim for a wider field of upward vision
  • Provides protection even in extremely low temperatures
Size: 510 to 610 mm
Certifikace: EN 397