Tisk…
uvex i-5 Safety Spectacles
Dodavatel: UVEX
9183.281 9183.265 9183064
111-2448EA 877 CZK
111-2448 111-2449 UVEX9183064
uvex i-5 Safety Spectacles
Brýle
Reliable protection thanks to an innovative and modern design. With an x-tended eyeshield and sideshield, the uvex i-5 can be relied on to protect the eyes in any situation. Sophisticated features ensure superior wearer comfort, even during long periods of use.
  • Soft, flexible nose pad for maximum comfort and a non-slip fit
  • x-tended eyeshield provides additional protection from flying particles, dust, and dirt
  • x-tended sideshield for increased side protection
  • 3-position side arm inclination adapts easily to any facial shape
  • Reliable UV 400 protection
More Product Information