Tisk…
Reusable earplugs, uvex whisper
Dodavatel: UVEX
2111.201 2111.237
UVEX2111.201EA 3990 CZK
UVEX2111.201 UVEX2111.237
Reusable earplugs, uvex whisper
Chrániče zátkové Chrániče zátkové k opakovatelnému použití
The earplugs with cord conveniently are at hand, whenever required. Manufactured from dirt repellent materials these ear plugs are suitable for use in dirty working environments.
  • With cord; can be worn comfortably around the neck
  • Hygienic storage in an individual carrying case
  • Dirt repellent surface can be cleaned using soap and water
  • Maintain the wearer's comfort and safety
Suitable for low noise areas.
SNR: 23 dB
Certifikace: EN 352-2:2002