Tisk…

Anti-VCP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: US Biological
V1035-20E-HRP
USBIV1035-20E-HRPEA 25170 CZK
USBIV1035-20E-HRP
Anti-VCP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: VCP (valosin containing protein)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu VCP
Environmentally Preferable
Název antigenu Valosin containing protein
Druh protilátky Primární
Klonovalita Polyclonal
Spojení HRP (Horseradish Peroxidase)
Reaktivita Human
Hostitel Rabbit
Izotyp IgG
Western blot Yes
ELISA Yes
Epitop C-terminal
Formulář liquid
Čištění purified by immunoaffinity chromatography
Skladovací pufr PBS, pH 7,2
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR