Tisk…

Anti-ULK1 Rabbit Polyclonal Antibody

Dodavatel: US Biological
U1500-70B
USBIU1500-70BEA 19000 CZK
USBIU1500-70B
Anti-ULK1 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: ULK1 (unc-51 like autophagy activating kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR