Tisk…

Anti-UBE2M Rabbit Monoclonal Antibody [clone: D13D7]

Dodavatel: US Biological
U1000-91E
USBIU1000-91EEA 17290 CZK
USBIU1000-91E
Anti-UBE2M Rabbit Monoclonal Antibody [clone: D13D7]
Protilátky
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: UBE2M (ubiquitin-conjugating enzyme E2M)
Clonality: monoclonal
Clone: D13D7
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR