Tisk…

Anti-TH Rabbit Polyclonal Antibody

Dodavatel: US Biological
T9237-10K
USBIT9237-10KEA 19110 CZK
USBIT9237-10K
Anti-TH Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Immunofluorescence: 1:100; Western Blot: 1:1000; Immunochemistry: 1:100; Immunoprecipitation: 1:100

Type: Primary
Antigen: TH (tyrosine hydroxylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR