Tisk…

Anti-TIA1 Goat Polyclonal Antibody

Dodavatel: US Biological
T5461-95B
USBIT5461-95BEA 19390 CZK
USBIT5461-95B
Anti-TIA1 Goat Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:128000; Western Blot: 0.1-0. 3?g/ml; Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: TIA1 (TIA1 cytotoxic granule-associated RNA binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR